VITLAB® Wide-Mouth Reagent Bottles, PP with Screw Caps, PP, BrandTech
Supplier: BrandTech
V101989 V101689 V101589 V101889 V101789
83008-552PK 351.91 CAD
83008-552 83008-540 83008-544 83008-536 83008-548
VITLAB® Wide-Mouth Reagent Bottles, PP with Screw Caps, PP, BrandTech
Bottles Roller Bottles
Slope-shouldered polypropylene bottles feature polypropylene screw caps.They provide excellent chemical resistance to aliphatic alcohols, aldehydes, aliphatic hydrocarbons, alkalis, and acids.
  • Bottles and caps are autoclavable
For replacement caps, see 83009-032 series.