Leak-Resistant Bottles, Amber, High-Density Polyethylene, Wide Mouth, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
209625 209626 209627 209628 209629
16155-625CS 506.8 CAD
16155-625 16155-626 16155-627 16155-628 16155-629
Leak-Resistant Bottles, Amber, High-Density Polyethylene, Wide Mouth, Wheaton®, DWK Life Sciences
Bottles Plastic Bottles
Ideal storage bottles. Designed with a special pour lip to facilitate safe, clean dispensing.
  • Amber in color to protect light-sensitive samples
  • Wide mouth design facilitates easy filling and pouring
  • Double-seal closure for leak-resistance
Ordering information: Supplied in convenience-packed cartons with polypropylene caps attached.