Chemware® Teflon® Resin PFA Jars, Saint-Gobain
Supplier: Saint Gobain Performance Plastics
D1069024 D1069025 D1069027 D1069029 D1069031 D1069706
36324-000EA 202.73 CAD
36324-000 36324-001 36324-002 36324-003 36324-004 36324-005 36324-006
Chemware® Teflon® Resin PFA Jars, Saint-Gobain
Bottles Plastic Bottles
Store solutions, pastes, and bulk materials without leaching, corrosion, contamination, or evaporation.
  • Knurled top for sure grip. Large mouth for easy access
  • Secondary inner seal for safe storage of any substance
  • Heat to 260 °C (500 °F). Freeze to –195 °C (–320 °F)
  • Stackable, easily cleaned
Certifications: Jar 36324-007 meets EPA requirement for soil sampling.