Jar, Polystyrene, Clear, Wide Mouth
Supplier: Qorpak
PLA-03357 PLA-03360 PLA-03359 PLA-03361 PLC-03730 PLC-03729 PLC-03727 PLC-03726 PLC-03724 PLC-08748 PLA-03359
16087-160CS 305.67 CAD
16087-160 16087-161 16087-167 16087-169 36318-862 36318-864 36318-866 36318-868 36318-870 89210-130 16087-166
Jar, Polystyrene, Clear, Wide Mouth
Bottles Plastic Bottles
Rigid, clear polystyrene jars offer glass-like clarity with a smooth surface that is slip-resistant.
  • Jars are ideal for storing and collecting specimens and lab supplies
  • Wide neck and straight sides facilitate easy recovery of contents