Chemware® Bottles, Teflon® PFA, Wide Mouth, Saint-Gobain
Supplier: Saint Gobain Performance Plastics
D1069591 D1069592 D1069593 D1069590
16128-539EA 130.52 CAD
16128-539 16128-541 16128-542 16128-543 16128-540
Chemware® Bottles, Teflon® PFA, Wide Mouth, Saint-Gobain
Bottles Plastic Bottles
Good for trace analysis. Leakproof, unbreakable PFA bottles for storage of virtually all corrosive chemicals including HF, nitric and perchloric acids.
  • Bottles withstand temperatures from 500 °F (260 °C) to the cryogenic range without becoming brittle
  • Chemically inert, non-stick surface is easy to clean
  • PFA cap