Leak-Resistant Bottles, High-Density Polyethylene, Wide Mouth, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
209550 209545 209546 209547 209548 209549
16153-513CS 806.4 CAD
16153-513 16155-545 16155-546 16155-547 16155-548 16155-549
Leak-Resistant Bottles, High-Density Polyethylene, Wide Mouth, Wheaton®, DWK Life Sciences
Bottles Plastic Bottles
Durable bottles are ideal for general laboratory use and field sample analysis.
  • Wide mouth design facilitates easy filling and pouring
  • Designed with a special pour lip to facilitate safe, clean dispensing
  • Double-seal closure for leak-resistance
Ordering information: Supplied in convenience-packed cartons with polypropylene caps attached.
Order Now
SPECIFICATIONS
Environmentally Preferable