SP Bel-Art Wide Mouth Storage Bottles, Heavy-Duty Closure, LDPE, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F10906-0500 F10906-1000 F10906-2000 F10906-0010 F10906-0050 F10906-0100 F10906-0200 F10906-0250 F10906-5000 F10906-1500
16126-278PK 150.76 CAD
16126-278 16126-314 16126-358 47750-656 16126-176 16126-198 16126-212 16126-234 70680-364 70680-366
SP Bel-Art Wide Mouth Storage Bottles, Heavy-Duty Closure, LDPE, Bel-Art Products, a part of SP
Bottles Roller Bottles
Low-density polyethylene bottles with a heavy-threaded, self-sealing screw cap closure provide reliability when storing, shipping, and handling materials in both liquid and powder form.
  • LDPE construction
  • Heavy-threaded, self-sealing screw cap closure
  • Reliable for use with both liquid and powder contents
  • Larger sizes are made of HDPE
Note: 5000 and 10000 ml sizes are made of high-density polyethylene and have an octagonal shape. They are not leakproof and are best used for dry materials.