Calcein AM fluorescent dye, Corning®

Supplier: Corning

354216 354217
CA89044-502EA 346.08 CAD
CA89044-502 CA89044-504
Calcein AM fluorescent dye, Corning®
Calcein AM

The fluorescent dye calcein acetoxymethylester (calcein AM) can be used to label cells when performing analyses such as tumour cell invasion, endothelial cell migration, endothelial cell tubulogenesis, and other cell-based assays.


  • Non fluorescent cell viability indicator
  • Cell permeant compound that is hydrolysed by intracellular esterases in the fluorescent anion calcein


Calcein AM can be used to fluorescently pre- and post-label viable cells and perform kinetic and endpoint experiments, respectively.


The fluorescent dyes, calcein acetoxymethylester (calcein AM), can be used with Falcon* Corning FluoroBlok* cell culture inserts and insert systems to label cells when performing analyses such as tumor cell invasion, endothelial cell migration, and other cell-based assays


Caution: NOTE: Cells are known to tolerate different fluorescent dyes for varying amounts of time in culture. Cells labelled with calcein AM should be exposed to the dye for less than eight hours.

Formula: C₄₆H₄₆N₂O₂₃
MDL Number: MFCD05861516
CAS Number: 148504-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR