Nalgene® Large Bottles, Polypropylene, Wide Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
2121-0005 2121-0010
16129-083CS 248.66 CAD
16129-083 16129-095
Nalgene® Large Bottles, Polypropylene, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Specifically designed for applications requiring autoclaving of large volumes.
  • Not recommended for shipping liquids
  • Supplied with 100-415 polypropylene closure
  • Not leakproof
  • Autoclavable
Note: Before autoclaving, set closure on top of bottle without engaging threads.