Imperial™ Protein Stain, Thermo Scientific
Supplier: Thermo Scientific
24615 24617
CAPI24615EA 354.56 CAD
CAPI24615 CAPI24617
Imperial™ Protein Stain, Thermo Scientific
Stains Electrophoresis Stains
Imperial Protein Stain is a ready-to-use colorimetric stain formulated with coomassie dye R-250 that delivers consistent nanogram-level detection of proteins in polyacrylamide electrophoresis gels or nitrocellulose membranes.
  • Sensitive—detects less than 3 ng protein per band with the enhanced protocol (3 hours)
  • Time to stain—ready-to-use reagent detects less than 6 ng protein per band in just 20 minutes
  • High contrast—intense purple bands are easier to photograph or scan than typical coomassie blue stains
  • Compatibility—compatible with downstream mass spectrometry analysis and protein sequencing
  • Additional reagents—water washes only; no acid-fixative or methanol destaining solutions required
  • Stable—1-year room-temperature stability
The stain is a unique formulation of coomassie brilliant blue R-250 that delivers substantial improvements in protein-staining performance compared to homemade or other commercial stains. Staining results in intensely colored protein bands that are easy to photograph and document with gel imagers. This reagent is one of the most sensitive colorimetric stains available, easily detecting 3 to 6 nanograms of protein per band. The easy-to-follow protocol is flexible to meet demanding time and sensitivity requirements.
Caution: Upon receipt store product at room temperature. For Research Use Only. Not for use in diagnostic procedures.