Propidium Iodide, Calbiochem®

Supplier: MilliporeSigma
Warning

Synonyms: 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide

537059-100MG 537059-1GM 537059-250MG 537059-50MG
CA80057-364EA 688.04 CAD
CA80057-364 CA80057-366 CA80057-368 CA80057-370
Propidium Iodide, Calbiochem®
Propidium iodide

Membrane-impermeable DNA intercalator. Has red fluorescence at 488nm. Useful for flow cytometry for staining apoptotic cells and nuclei. Can be used to differentiate between apoptotic and necrotic cell death. Can be used together with common green fluorescent probes labeled with fluorescein isothiocyanate.


Extinction coefficient (493nm): 5900M-1cm-1; (287nm): 55,700M-1cm-1. Excitation maximum: 536nm. Emission maximum: 617nm.

Formula: C₂₇H₃₄I₂N₄
MW: 668.4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011921
CAS Number: 25535-16-4
EINECS: 247-081-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR