Nalgene® Heavy-Duty Vacuum Bottles, Polypropylene Copolymer, Wide Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS2126-0250 2126-1000 2126-2000 2126-5000 2126-4000
16056-970CS 226.26 CAD
16056-970 16210-076 16210-078 16211-080 16331-100
Nalgene® Heavy-Duty Vacuum Bottles, Polypropylene Copolymer, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Sturdy, thick-walled bottles are designed for heavy-duty applications in the biotechnology lab.
  • Rugged caps with TPE gaskets guarantee leakproof service under vacuum
  • Autoclavable
Bottles withstand repeated application of full vacuum for 24-hour periods at room temperature (20 °C, 68 °F). Use as waste aspirator bottle (with Barbed Bulkhead Fittings 16225-232) or as a vessel for scale-up activities in cell culture applications.
Note: Before autoclaving, set cap on top of bottle without engaging threads.
Ordering information: For replacement caps, see 16219-122 series.