Specimen Zipper Bags with Pouch and Biohazard Message, Associated Bag
Supplier: Associated Bag
14-90 14-93
89085-394CS 259.29 CAD
89085-394 89085-396
Specimen Zipper Bags with Pouch and Biohazard Message, Associated Bag
Bags Waste Disposal Bags
Sure-seal zipper bag holds specimen and protects against accidental spills.
Separate document pouch keeps documents with specimens during transport and storage. Meets current labeling requirements of OSHA standard 29CFR1910.1030 regarding biohazard symbols.