VentSep® Separatory Funnels, Pear-Shape, Glas-Col®
Supplier: Glas-Col
099ASST555 099ASST655
57020-300EA 1954.89 CAD
57020-300 57020-301
VentSep® Separatory Funnels, Pear-Shape, Glas-Col®
Funnels Separating Funnels
These separatory funnels feature a built-in relief valve, a fluoropolymer resin stopcock, and a screw cap with fluoropolymer resin liner
Funnels vent gas pressure over 1psi automatically. Funnels are ideal for vigorous hand or machine shaking.