Nalgene® Separatory Funnels, Polypropylene, Squibb, Pear-Shape, Thermo Scientific
Supplier: Thermo Fisher Scientific
4300-0125 4300-0250 4300-0500 4300-1000
30356-744CS 1376.39 CAD
30356-744 30356-766 30356-788 30356-802
Nalgene® Separatory Funnels, Polypropylene, Squibb, Pear-Shape, Thermo Scientific
Funnels Separating Funnels
Translucent, leakproof funnels feature excellent chemical resistance and do not require lubrication
Compatible with a wide range of lab reagents. Separation lines between immiscible liquids are easy to observe. Funnel and screw closure are autoclavable; stopcock assembly may be chemically disinfected.