VITLAB® Graduated Cylinders, Polypropylene, Class B, Blue Scale, BrandTech
Supplier: Brandtech
646081 648081 651081 653081 647081 649081 650081 652081
83008-860PK 212.9 CAD
83008-860 83008-880 83008-910 83008-930 83008-870 83008-890 83008-900 83008-920
VITLAB® Graduated Cylinders, Polypropylene, Class B, Blue Scale, BrandTech
Cylinders Measuring Cylinders
Made of polypropylene with ring-molded graduations overprinted with blue screening to simplify measurements.
  • Translucent polypropylene is chemically resistant
  • Blue screening for high contrast with many liquids
These VITLAB® graduated cylinders are made of polypropylene for durability and chemical resistance. Cylinders are highly translucent with molded graduations, and blue-screened overprint for easy meniscus readings. Hexagonal base resists rolling.
Certifications: Class B per DIN 12681.