VITLAB® Graduated Cylinders, PMP, Class A, Certified, BrandTech
Supplier: Brandtech
64804 64704 64904 65004 65204 65304 64604 65104
83008-878PK 73.37 CAD
83008-878 83008-868 83008-888 83008-898 83008-908 83008-918 83008-928 10117-442 83008-909
VITLAB® Graduated Cylinders, PMP, Class A, Certified, BrandTech
Cylinders Measuring Cylinders
Made of polymethylpentene (PMP) with ring-molded graduations at major increments for excellent readability
  • Glass clear PMP
  • Certificate supplied
Graduated at increments of 1 or 2% of nominal volume for precise measuring. Cylinders are marked with 'H', lot number, and year of production.
Certifications: Class A per DIN 12681 with test certificate.
Order Now
SPECIFICATIONS
Form Short
Environmentally Preferable