Cylinder Bottles, PVC, Narrow Mouth, Qorpak®
Supplier: Qorpak
PLA-05756 PLA-05758 PLA-05759 PLC-05772
16087-414CS 677.92 CAD
16087-414 16087-417 75877-328 76470-978 76470-980 76470-982
Cylinder Bottles, PVC, Narrow Mouth, Qorpak®
Bottles Plastic Bottles
Clear PVC Cylinder Bottles provide an excellent resistance to oxygen permeation, oils and good water barrier characteristics.
  • Transparent, rigid cylinders allow for easy visual identification of the amount of product inside
Narrow mouth cylinders are perfect for handling liquid samples in the lab. The shoulder style allows you to pour small quantities without spilling.