VWR® Traceable® Alarm Thermometer/Alarm Timer
Supplier: VWR
VWR®
4147
62344-916EA 155.16 CAD
62344-916
VWR® Traceable® Alarm Thermometer/Alarm Timer
Thermometers Digital Thermometers
Unit is ideal for monitoring liquids, air/gas, or semi-solids in water baths, ovens, refrigerators, incubators, and tanks.
  • Traceable®
  • Reads °C and °F
  • Easy-to-read display
  • Chemical-resistant ABS base
Easy-to-read “pop-up” display shows time remaining, probe temperature, and temperature alarm setting
Timer counts down from 23 hours, 59 minutes, 59 seconds in one-minute increments. Alarm sounds when time reaches zero. LCD flashes a times-up message and timer begins counting up. For repetitive applications, memory returns time to previous programmed setting.
Fold-back display panel and magnets on the back allow for vertical mounting. Base is made of high-impact, chemical-resistant ABS.
Dual alarms sound separately for programmed time and temperature settings. Five-minute alarm sounds when temperature reading rises above programmed setting. Alarm continues to sound until switched off or temperature returns to an “in-range” condition.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a detachable 21.6 cm (81/2") curved probe with 107 cm (42") long cable and a replaceable AAA alkaline battery.