Graduated Cylinders, PMP, Kartell
Supplier: VWR International
24774-604 24774-637 24774-660 24774-692 24774-728 24774-750 24774-783 24774-785
24774-604CS 452.79 CAD
24774-604 24774-637 24774-660 24774-692 24774-728 24774-750 24774-783 24774-785
Graduated Cylinders, PMP, Kartell
Cylinders Measuring Cylinders
These polymethylpentene cylinders are transparent, shatterproof, and autoclavable.
  • Transparent
  • Shatterproof
  • Autoclavable
  • Polymethylpentene fabrication
Graduated cylinders withstand temperatures of 0 to 120°C (32 to 248°F) when used continuously, and withstand temperatures as high as 170°C (338°F) when used for short periods of time. Permanent graduations molded in. Meet stringent international quality criteria according to ISO 6706 certification. Not resistant to strong oxidizers, some hydrocarbons, and chlorinated solvents.
Order Now
SPECIFICATIONS
Form Short
Environmentally Preferable