PYREX® Double Metric Scale Graduated Cylinders, Class A, To Deliver, Corning
Supplier: Corning
3023-1L 3023-2L 3026-1L 3026-2L 3023-100 3026-100 3023-250 3026-250 3023-25 3026-25 3023-500 3026-500 3023-50 3026-50
22877-030CS 228.08 CAD
22877-030 22877-032 22877-150 22877-152 89090-488 89090-490 89090-492 89090-494 89090-496 89090-498 89090-500 89090-502 89090-504 89090-506
PYREX® Double Metric Scale Graduated Cylinders, Class A, To Deliver, Corning
Cylinders Measuring Cylinders
PYREX® borosilicate glass, tall form, class A.
  • Calibrated to deliver (TD) in accordance with ASTM E-1272 and ASTM E-542
  • Double metric scale is numbered both up and down in durable white or blue enamel
  • Hexagonal base and pourout
  • With shock absorbing polyethylene bumper guards