PYREX® VISTA™ Graduated Cylinders, Class A, To Deliver, Corning
Supplier: Corning
70024-100 70024-10 70024-250 70024-25 70024-500 70024-50
89090-446CS 602.46 CAD
89090-446 89090-450 89090-454 89090-458 89090-462 89090-466
PYREX® VISTA™ Graduated Cylinders, Class A, To Deliver, Corning
Cylinders Measuring Cylinders
PYREX® borosilicate glass, class A.
  • Calibrated to deliver (TD) in accordance with ASTM E-1272
  • Hexagonal base with graduations in blue enamel
  • 10 ml cylinder has a funnel top