PYREX® Graduated Cylinder, To Contain, with PYREX® Stopper, Corning
Supplier: Corning
2982-1L 2982-2L 2982-100 2982-10 2982-250 2982-25 2982-500 2982-50
24760-167CS 347.61 CAD
24760-167 24760-189 89090-620 89090-632 89090-800 89090-830 89090-886 89090-924
PYREX® Graduated Cylinder, To Contain, with PYREX® Stopper, Corning
Cylinders Measuring Cylinders
Borosilicate glass with hollow glass tapered stopper.
  • Calibrated to contain (TC, In)
  • Hexagonal base
  • Ring marks at major graduations, marks and inscriptions in durable white enamel