PYREX® Graduated Cylinders, To Deliver, Corning
Supplier: Corning
3024-2L 3024-4L 3024-PACK 3024-1L 3024-100 3024-10 3024-250 3024-25 3024-500 3024-50 3024-5
24709-782CS 368.94 CAD
24709-782 32645-066 77777-014 24709-771 89090-554 89090-558 89090-562 89090-566 89090-570 89090-572 89090-574
PYREX® Graduated Cylinders, To Deliver, Corning
Cylinders Measuring Cylinders
Hexagonal base with white enamel metric scale.
Sizes 5 and 10 ml are funnel top. Sizes 25 to 2000 ml have bumper guard.
Ordering information: The variety pack (77777-014) includes one of each of the following cylinders: 10, 25, 50, 100, and 250 ml.