NIGHTSEA® White Light Head
Supplier: Electron Microscopy Sciences
SFA-LH-WH SFA-LH-WH
77463-326EA 770.55 CAD
77463-326 CA77463-326
NIGHTSEA® White Light Head
Lights Fluorescent Lights, Lamps and Bulbs
Use as a convenient option for general illumination and as a focusing aid for fluorescence imaging. Just exchange the fluorescence excitation light head module for the white-light module in a matter of seconds.
  • Compatible with the NIGHTSEA® SFA stereo microscope fluorescence adapter system
An extremely convenient option for general illumination and as a focusing aid for fluorescence imaging. When using the NIGHTSEA® system for fluorescence, there is no need for a separate white light source. Just exchange the fluorescence excitation light head module for the white-light module in a matter of seconds.