VITLAB® Square Laboratory Bottles, HDPE, Narrow Mouth, with PP Screw Caps, BrandTech
Supplier: BrandTech
VITLAB®
V92189 V92089 V91789 V91989
83009-054PK 331.36 CAD
83009-054 83009-050 83009-052 83009-053
VITLAB® Square Laboratory Bottles, HDPE, Narrow Mouth, with PP Screw Caps, BrandTech
Bottles Plastic Bottles
General-use, high-density polyethylene laboratory bottles come with polypropylene screw caps.
  • Space-saving shape
  • HDPE construction with PP screw caps
  • Narrow mouth
Bottles provide excellent chemical resistance to aliphatic and aromatic hydrocarbons, alcohols, aldehydes, alkalis, and acids.