Nalgene® Square Laboratory Bottles, Polypropylene Copolymer, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
2016-0030 2016-0060 2016-0125 2016-0250 2016-0500 2016-1000
16120-732CS 262.91 CAD
16120-732 16120-737 16120-741 16120-745 16120-749 16120-753
Nalgene® Square Laboratory Bottles, Polypropylene Copolymer, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Square laboratory bottles provide excellent chemical resistance.
  • Leakproof
  • Repeatedly autoclavable
Note: Before autoclaving, set closure on top of the bottle without engaging the threads.
The translucent design is ideal for long-term storage of inorganic buffers, alcohols, acids, and bases. All sizes have permanent graduations in convenient increments. Bottles are designed for use within a temperature range of 0 to 135 °C (32 to 275 °F).
Ordering information: Supplied with polypropylene screw closures assembled on bottles. For replacement capsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable