Staticide® Anti-Static Neutralizer/Cleaner, ACL Staticide
Supplier: ACL Staticide
Staticide®
4020-2 4020-5 4020-1
21831-044EA 5342.26 CAD
21831-044 21831-042 21831-046
Staticide® Anti-Static Neutralizer/Cleaner, ACL Staticide
Surface Treatments Floor Treatments
A highly concentrated, biodegradable, antistatic cleaner that leaves no film or streaks and will not degrade a production environment’s antistatic properties
  • Exhibits a low foam level and constant detergency
  • The formulation contains no phosphates or alkalis and is safe for septic and sewage systems
Static charge decay: 0.2 s
Certifications: Meets MIL-B-81705B, NFPA 56A. Electrostatic decay time is measured per FED Test Method 4046.