Nalgene® High-Density Polyethylene Bottles, Sterile, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
342002-9025 342002-9050 342089-0001 342089-0002 342089-0004 342089-0008 342089-0016 342089-0032
73520-062CS 1056.7 CAD
73520-062 73520-064 73520-066 73520-068 73520-070 73520-072 73520-074 73520-076
Nalgene® High-Density Polyethylene Bottles, Sterile, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
HDPE bottles feature a narrow mouth and leakproof closure design.
Manufactured and packaged in controlled environments to minimize biological and particulate contamination. Gamma irradiated to 10⁻⁶ sterility assurance levels.
Certifications: Bottles meet USP-VI and are noncytotoxic and nonpyrogenic.
Ordering information: Supplied with white polypropylene closures.