Nalgene® Buchner Funnels, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
4280-0550 4280-0700 4280-0900 4280-1100 4280-1500
30305-040CS 223.42 CAD
30305-040 30305-062 30305-084 30305-109 30305-120
Nalgene® Buchner Funnels, Polypropylene, Thermo Scientific
Funnels Filtration Funnels Büchner Funnels
Sturdy two-piece construction
Separable for easier cleaning. Highly resistant to most chemicals. Autoclavable.