Nalgene® Low-Particulate Bottles, High-Density Polyethylene, Narrow Mouth, with Screw Caps, Thermo Scientific
Supplier: Thermo Fisher Scientific
382099-0250 382099-0500 382099-1000
16060-072CS 434.82 CAD
16060-072 16060-074 16060-076
Nalgene® Low-Particulate Bottles, High-Density Polyethylene, Narrow Mouth, with Screw Caps, Thermo Scientific
Bottles Plastic Bottles
Packaging/shipping bottles are ideal for microelectronic reagents and biotech/pharmaceutical applications.
  • Leakproof to 15 psi
High-density polyethylene containers have an average particle level of less than 30 particles per ml at 0.3 µm and greater.
Ordering information: Complete with polypropylene screw caps. Certificate of quality included with each case.
Order Now
SPECIFICATIONS
Environmentally Preferable