Signature Series™ Clarifier™ 1, 2, Epredia
Supplier: RICHARD-ALLAN SCIENTIFIC CA
7401 7402 7402L
CA41810-468CS 140 CAD
CA41810-468 CA41810-474 CA89237-218
Signature Series™ Clarifier™ 1, 2, Epredia
Stains Microscopy and staining reagents
Enhance the nuclear definition and clarity of Signature Series hematoxylins with Epredia™ Signature Series Clarifier™ 1, 2.
  • Improves contrast between Nuclei and Cytoplasm
  • Increases cell transparency
  • Removes background staining caused by excessive tissue adhesive
  • Removes unwanted hematoxylin staining from adhesive without affecting nuclear staining
Designed to remove background stain by excessive use of tissue adhesive and to increase transparency. Both Clarifier 1 and 2 selectively remove hematoxylin staining from excess adhesive without affecting nuclear staining. Use of Clarifier 1 or 2 improves contrast between nuclei and cytoplasm.
Use Thermo Scientific™ Richard-Allan Scientific™ Modified Harris Hematoxylin, a regressive nuclear stain, for both histology and cytology. Produce crisp nuclear staining with Thermo Scientific™ Richard-Allan Scientific™ Modified Mayer's Hematoxylin, a progressive nuclear stain. Enhance the nuclear definition and clarity of Signature Series hematoxylins with Thermo Scientific™ Richard-Allan Scientific™ Signature Series Clarifier™ 1, 2.
Produce very dense color saturation with Thermo Scientific™ Richard-Allan Scientific™ May-Grunwald Stain, a classical hematology stain. Identify immature erythrocytes with Thermo Scientific™ Richard-Allan Scientific™ Reticulocyte Stain, a specialized hematology stain.With three strengths available, the Thermo Scientific™ Richard-Allan Scientific™ Gill™ 1, 2, 3 Hematoxylin allows complete control of staining intensities.
Control the color hues and intensities of your work with Thermo Scientific™ Richard-Allan Scientific™ Papanicolaou Stain EAs, useful for both gynecological and non-gynecological staining. Stain gynecological specimens with Thermo Scientific™ Richard-Allan Scientific™ Papanicolaou Stain OGs. Achieve results similar to conventional Papanicolaou stains with Thermo Scientific™ Richard-Allan Scientific™ Signature Series Cyto-Stain™, a uniquely formulated cytoplasmic stain. Create an ideal visual contrast to Hematoxylin with the fast-acting Thermo Scientific™ Richard-Allan Scientific™ Signature Series Eosin-Y counterstain. Offers excellent delineation of cytoplasmic components. Yield visually similar results to Gills 1 and 2 with Thermo Scientific™ Richard-Allan Scientific™ Signature Series Hematoxylin 1, a uniquely formulated product.
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Thermo Scientific™ Richard-Allan Scientific™ Modified Grocott's Methenamine Silver (Periodic Acid), a uniquely modified silver stain formulation that utilizes methenamine/borax capsules to make working silver solution.