VWR® Narrow Mouth Laboratory Bottles, Amber HDPE
Supplier: VWR International
414004-160 414004-161 414004-162 414004-163 414004-164 414004-165 414004-166 414004-167 414004-168 89221-612 89221-614 89221-616 89221-618 89221-620 89221-622 89221-624 89221-626 89221-628
414004-160CS 216.36 CAD
414004-160 414004-161 414004-162 414004-163 414004-164 414004-165 414004-166 414004-167 414004-168 89221-612 89221-614 89221-616 89221-618 89221-620 89221-622 89221-624 89221-626 89221-628
VWR® Narrow Mouth Laboratory Bottles, Amber HDPE
Bottles Plastic Bottles
Bottles are for general laboratory use and for storing, shipping, and packing liquids, especially light-sensitive materials or photosensitive samples.
  • Amber coloring protects light-sensitive contents
  • High-density polyethylene fabrication
  • Excellent chemical resistance
Can be stored in freezer to −100 °C (−148 °F), and have excellent chemical resistance to most acids, bases, and alcohols. Amber polypropylene screw closure included. Bottles comply with current USP requirements for maximum light transmission.