Pre-Cleaned Bottles, HDPE, Narrow Mouth, Environmental Express®
Supplier: Environmental Express
BPC3010 BPC3020 BPC3000 BPC1020 BPC1000 BPC1030 BPC1001 BPC3001 BPC1011 BPC3011 BPC3021 BPC1010 BPC1021
23050-142CS 312.22 CAD
23050-142 23050-148 23050-154 23050-158 23050-250 23050-300 23050-350 23050-352 23050-354 23050-356 23050-360 23050-130 23050-359
Pre-Cleaned Bottles, HDPE, Narrow Mouth, Environmental Express®
Bottles Plastic Bottles
All containers meet one of three levels of pre-cleaning.
These high-density polyethylene bottles include polypropylene closures with F-217 PE liners.
Level III indicates that containers have not received an EPA washing treatment. Level II indicates that each case of containers has been custody sealed and lot number labeled for traceability to the enclosed Statement of Compliance. Level I pre-cleaning is the same as Level II, but instead of a Statement of Compliance, a Certificate of Analysis is provided indicating that the containers received full EPA quality assurance treatment.