Graduated Addition Funnel, Fluoropolymer Resin Stopcock, Chemglass
Supplier: Chemglass
CG-1712-03 CG-1712-06 CG-1712-01 CG-1712-02 CG-1712-04 CG-1712-10 CG-1712-13 CG-1712-16 CG-1712-19
80070-640EA 864.88 CAD
80070-640 80070-646 80070-636 80070-638 80070-642 80070-654 80070-660 80070-666 80070-672
Graduated Addition Funnel, Fluoropolymer Resin Stopcock, Chemglass
Funnels Dropping Funnels
Cylindrical style addition funnel graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line
With a [ST] outer joint and lower inner joint, both the same size, and pressure equalizing arm.