SP Bel-Art Precisionware® Narrow Mouth Bottles, Autoclavable Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F10631-0007 F10631-0008
74950-013PK 212.49 CAD
74950-013 74950-015
SP Bel-Art Precisionware® Narrow Mouth Bottles, Autoclavable Polypropylene, Bel-Art Products, a part of SP
Bottles Plastic Bottles
Ideal for pharmaceutical or similar packaging, polypropylene Boston round bottles are clear and molded from FDA compliant material.
  • Polypropylene construction
  • Ribbed cap exteriors
  • Autoclavable, with caps detached, to 121 °C (250 °F)
Polypropylene caps have a valve-seal design for a leakproof fit; deep square threads help prevent closure back-off during shipping. Ribbed cap exteriors are designed for torque closure and easy opening. Inner bottle surfaces are smooth and rounded for easy cleaning and thorough emptying.