VITLAB® Reagent Bottles, PFA, Narrow Mouth, BrandTech
Supplier: Brandtech
108297 108397 108497 109297 109397
83008-588PK 678.68 CAD
83008-588 83008-592 83008-596 83008-604 83008-606
VITLAB® Reagent Bottles, PFA, Narrow Mouth, BrandTech
Bottles Plastic Bottles
Leach-resistant bottles are for long-term storage of solvents and standards.
  • Bottles are made of perfluoroalkoxy copolymer (PFA)
  • Highly resistant to heat and chemical contamination
  • PFA’s smooth surface and hydrophobic
  • Anti-adhesive properties simplify the cleaning process, making it ideal for trace analysis
  • Bottles and caps are autoclavable
Bottles maintain stability in temperatures from –200 to 260 °C (–328 to 500 °F). Thread finish: S 28.
Delivery information: Supplied with PFA screw cap with a molded sealing ring.