SAFEGUARD® Specimen Bags
Supplier: Minigrip
MGTZ69 MGTZ810 MGTZ1215
89005-356CS 213.84 CAD
89005-356 89005-360 89005-362
SAFEGUARD® Specimen Bags
Bags Sample Bags
Reclosable three-wall specimen transport bags feature distinct black print on an orange background, with biohazard logo and check boxes labeled Freeze, Refrigerate, and Room Temperature.
Bags meet NCCLS M29-T2 specimen handling guidelines and OSHA 1910.1030 regulation for blood-borne pathogens containment and biohazard graphics.
The contents are accessible through a unique TearZone® feature that allows the bag to be opened without perforations that may compromise the safety of the bag or the health of the technician. Outside pouch provides protection and transportation for patientMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable