VWR® Disposable Funnels
Supplier: VWR International
414004-288 414004-289 414004-290
414004-288CS 131.33 CAD
414004-288 414004-289 414004-290
VWR® Disposable Funnels
Funnels Filtration Funnels Filter Funnels
Funnels are semi-transparent and are provided in a variety of stem lengths for varied pouring purposes.
  • Tapered at a 60° angle
  • Inside fluting
  • Polystyrene or polypropylene construction
Disposable funnels are inexpensive and can be purchased in bulk packs.