Nalgene® PassPort™ IP2 Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
2099-0001 2099-0002 2099-0004 2099-0008 2099-0016 2099-0032 2099-0010
16057-160CS 212.35 CAD
16057-160 16057-162 16057-164 16057-166 16057-168 16057-169 16057-170
Nalgene® PassPort™ IP2 Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Excellent for small-scale inter-lab shipping of hazardous samples and chemicals, such as corrosives and cleaning compounds.
  • Leakproof
  • With polypropylene screw closures
The application of pressure or vacuum may result in product failure, damage to property, or personal injury.
Recommended for customers who are designing, assembling, and certifying their own combination packaging. Equipped with heavy-duty walls, bottles are designed to resist splitting and puncturing from shipping shocks; proven effective by industrial users. IP2 marking, material, volume, registration notch, and NALGENE name are molded into the bottle's bottom. Evaluated at 15 psi (103 kPa) per 49CFR 173.27 (c) (2) ICAO Technical Instructions Part 3-1; 1.1.6.1, and IATA Dangerous Goods Regulations Section 5.0.2.9.
Ordering information: For replacement caps for 16057-169 and -170, see 16219-122 series.
Caution: Do not use bottle under pressure or vacuum.