Nalgene® Laboratory Bottles, Teflon® PFA, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS1630-0001 DS1630-0002 1630-0004 1630-0008 1630-0016 1630-0032
16071-561CS 978.34 CAD
16071-561 16071-563 16071-564 16071-566 16071-568 16071-570
Nalgene® Laboratory Bottles, Teflon® PFA, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Teflon® PFA resists temperature, chemicals, and corrosion.
  • Withstand temperatures from –270 to 250 °C (–454 to 482 °F)
  • Translucent bottles allow liquid levels to be seen
  • Linerless, leakproof closures made of Teflon PFA for a positive seal
  • Leakproof, Autoclavable
Note: Before autoclaving, set closure on top of bottle without engaging threads.
Inert to virtually all chemicals except molten alkali metals, fluorine at elevated temperatures, and certain complex halogenated compounds at elevated temperatures and pressures.