Nalgene® Laboratory Bottles, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
2003-9025 2003-9050 2003-0001 2003-0002 2003-0004 2003-0008 2003-0016 2003-0032
16057-980CS 119.19 CAD
16057-980 16058-000 16058-021 16058-043 16058-065 16058-101 16058-112 16058-123 16058-145
Nalgene® Laboratory Bottles, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Leakproof, translucent, flexible bottles, with excellent impact resistance and better visibility to contents than HDPE bottles.
  • Excellent chemical resistance to most acids, bases, and alcohols
  • LDPE is extremely low in trace metal content
  • Excellent material for trace metal analysis
Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: For replacement closures for 16058-112 and 16058-145, see 16219-122 series.