Nalgene® Heavy-Duty Funnels, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
4260-0040 4260-0060 4260-0100
30250-109CS 383.23 CAD
30250-109 30250-062 30250-084
Nalgene® Heavy-Duty Funnels, Thermo Scientific
Funnels Filtration Funnels Filter Funnels
Funnels are completely molded, unbreakable and with vent-ribs for even flow
The largest size is molded of high-density linear polyethylene (HDPE). Other sizes are low density polyethylene (LDPE).