Nalgene® Utility Funnels, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
4256-0414 4256-0234 4256-0638
30255-306CS 2378.7 CAD
30255-306 30255-340 30255-383
Nalgene® Utility Funnels, Thermo Scientific
Funnels Filtration Funnels Filter Funnels
Useful in numerous laboratory applications
Funnel 30255-383 has a large throat opening for faster transfer of materials.
The three smaller sizes are molded of autoclavable polypropylene; the three larger sizes are high-density polyethylene.