PYREX® Fernbach Culture Flasks, Corning
Supplier: Corning
4423-2XL 4425-2XL 4426-2XL 4420-2XL
22877-168CS 1040.08 CAD
22877-168 22877-170 89090-136 89090-138 89091-278
PYREX® Fernbach Culture Flasks, Corning
Flasks Culture Flasks
Fernbach-style culture flasks are ideal for culturing organisms that require a large surface area-to-volume ratio, and in serum production
Baffled bottom flasks have three baffles at center on outside for maximal oxygen transfer to culture medium. Flasks that accept rubber stopper (not included) have heavily beaded rim to protect against chipping; also accept cotton plugs. Light blue phenolic screw caps are autoclavable and PTFE-lined. Orange plug seal cap is autoclavable and linerless, with drip-free pouring ring.
These culture flask are made of Pyrex® borosilicate glass.