KELKLAVE Autoclave Gloves, Wells Lamont
Supplier: Wells Lamont
422-11 422-5
80091-458EA 26.67 CAD
80091-458 80091-460
KELKLAVE Autoclave Gloves, Wells Lamont
Gloves
These autoclave gloves protect against temperatures up to 121 °C (250 °F).
  • White terry cloth construction
  • Clute cut and gauntlet cuff
Certifications: CE marked.