6×3 X-Gen Goggles
Supplier: UNIVET OPTICAL TECHNOLOGIES
MDU6003SC MDU6004SC
77393-766PK 201.45 CAD
77393-766 77393-768
6×3 X-Gen Goggles
Goggles
6×3 X-Gen goggles feature a panoramic lens that easily integrates with prescription glasses.
  • Lens treatment: anti-fog and anti-scratch (double coating)
  • Interchangeable lens system
  • Frame engineered for soft facial support
  • Compatible with filtering half masks
6×3 X-Gen goggles feature a lens with anti-fog and anti-scratch ultra performing Vanguard UDC treatment. They are certified according to EN166 standard, level 3 and 4 (EU) for protection against droplets, splashes, and large dust particles.
Ordering information: The 6×3 goggle face shield is available separately as a face protection accessory for this goggle series.