5×7 X-Gen Over Spectacles
Supplier: UNIVET OPTICAL TECHNOLOGIES
MDU5006S MDU5007SC MDU5032SC
77393-732PK 338.44 CAD
77393-732 77393-734 77393-752
5×7 X-Gen Over Spectacles
Safety Glasses
The 5×7 X-Gen over spectacles an be worn alone or over prescription glasses.
  • Temples with rubber terminals and tilting movement
  • Adjustable temple length
  • Integrated side shields
  • Multiple lens treatments are available, including anti-fog, anti-scratch, and UV525
Smoke lenses can help to reduce anxiety. UV525 protects eyes during curing light usage. VPlus anti-fog and anti-scratch enhancement is available on select models.