Sterilizable Overspec Goggles, 619 Polycarbonate
Supplier: KLARITEX NORTH AMERICA
619.04.23.10 619.05.23.10
77393-722CS 6446.4 CAD
77393-722 77393-724
Sterilizable Overspec Goggles, 619 Polycarbonate
Goggles
619 Polycarbonate sterilizable overspec goggles are available in direct vent or indirect vent formats.
  • Overspec goggles feature interchangeable lens technology
  • Meets EN 166 and EN 170 standards
  • Scale number: (2C-1.2)
  • Mechanical resistance: Medium energy impact (B)
  • Optical class (1)
Goggles with polycarbonate lens are reusable for up to 30 autoclave sterilization cycles with steam at 121 °C, 1 atm for 30 minutes. They are also reusable for up to 10 gamma irradiation cycles.