VWR® Wash Bottles
Supplier: VWR International
414004-225 414004-226 414004-227 414004-228
414004-225CS 362.09 CAD
414004-225 414004-226 414004-227 414004-228
VWR® Wash Bottles
Bottles Wash Bottles
LDPE wash bottle features a polypropylene screw cap and stem that are molded into a one-piece unit for leakproof operation.
  • LDPE construction
  • Provided with a polypropylene screw cap
  • Leakproof operation
Dispensing tube produces a fine stream, but can be cut for a coarser flow.
Order Now
SPECIFICATIONS
Environmentally Preferable